missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)benzamide, 97%

Catalog No. AAL0621209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0621209 10 g
AAL0621218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0621209 Supplier Thermo Scientific Chemicals Supplier No. L0621209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26385-07-9 | C9H10ClNO | 183.635 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26385-07-9
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.635
MDL Number MFCD00000964
InChI Key FYQJUYCGPLFWQR-UHFFFAOYSA-N
Synonym n-2-chloroethyl benzamide, benzamide, n-2-chloroethyl, acmc-1cb5c, 2-benzoylamino-ethyl chloride, fyqjuycgplfwqr-uhfffaoysa, n-2-chloroethyl benzamide 10g
PubChem CID 101439
IUPAC Name N-(2-chloroethyl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NCCCl

Specifications

Melting Point 101°C to 105°C
Quantity 10 g
Beilstein 1943939
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material N-(2-Chloroethyl)benzamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 247-641-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only