accessibility menu, dialog, popup

UserName

CAS RN 26385-07-9

IUPAC Name: N-(2-chloroethyl)benzamide
Synonyms: N-(2-chloroethyl)benzamide
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: FYQJUYCGPLFWQR-UHFFFAOYSA-N
SMILES: ClCCNC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results