missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)maleimide Hydrochloride 93.0+%, TCI America™

Catalog No. A2436200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2436200MG 200 mg
A24361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2436200MG Supplier TCI America Supplier No. A2436200MG

Chemical Identifiers

CAS 134272-64-3
Molecular Formula C6H9ClN2O2
Molecular Weight (g/mol) 176.6
MDL Number MFCD09800491
InChI Key NJQOCRDPGFWEKA-UHFFFAOYSA-N
PubChem CID 22118207
IUPAC Name 1-(2-aminoethyl)pyrrole-2,5-dione;hydrochloride
SMILES C1=CC(=O)N(C1=O)CCN.Cl

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 200 mg
Formula Weight 176.60
Percent Purity ≥93.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Aminoethyl)maleimide Hydrochloride