accessibility menu, dialog, popup

UserName

ONOH2NHCl

CAS RN 134272-64-3

IUPAC Name: 1-(2-aminoethyl)-2,5-dihydro-1H-pyrrole-2,5-dione hydrochloride
Synonyms: 1-(2-aminoethyl)pyrrole-2,5-dione hydrochloride
Molecular Weight (g/mol): 176.6
Molecular Formula: C6H9ClN2O2
InChi Key: NJQOCRDPGFWEKA-UHFFFAOYSA-N
SMILES: Cl.NCCN1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results