missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Aminoethyl)glycine 97.0+%, TCI America™
$246.05 - $1030.83
Chemical Identifiers
| CAS | 24123-14-6 |
|---|---|
| Molecular Formula | C4H10N2O2 |
| Molecular Weight (g/mol) | 118.14 |
| MDL Number | MFCD00144824 |
| InChI Key | PIINGYXNCHTJTF-UHFFFAOYSA-N |
| Synonym | Ethylenediamine-N-monoacetic Acid |
| PubChem CID | 428913 |
| IUPAC Name | 2-[(2-aminoethyl)amino]acetic acid |
| SMILES | NCCNCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1153100MG
|
TCI America
A1153100MG |
100 mg |
Each for $246.05
|
|
|||||
|
A11531G
|
TCI America
A11531G |
1 g |
Each for $1,030.83
|
|
|||||
Chemical Identifiers
| 24123-14-6 | |
| 118.14 | |
| PIINGYXNCHTJTF-UHFFFAOYSA-N | |
| 428913 | |
| NCCNCC(O)=O |
| C4H10N2O2 | |
| MFCD00144824 | |
| Ethylenediamine-N-monoacetic Acid | |
| 2-[(2-aminoethyl)amino]acetic acid |
Specifications
| 143°C | |
| White | |
| MFCD00144824 | |
| PIINGYXNCHTJTF-UHFFFAOYSA-N | |
| 2-[(2-aminoethyl)amino]acetic acid | |
| 118.14 | |
| 118.14 | |
| Crystalline Powder |
| 24123-14-6 | |
| C4H10N2O2 | |
| Ethylenediamine-N-monoacetic Acid | |
| NCCNCC(O)=O | |
| 100 mg | |
| 428913 | |
| ≥97.0% (T) | |
| N-(2-Aminoethyl)glycine |
Safety and Handling
TSCA : No