accessibility menu, dialog, popup

UserName

CAS RN 24123-14-6

H2NNHOHO

CAS RN 24123-14-6

IUPAC Name: 2-[(2-aminoethyl)amino]acetic acid
Synonyms: [(2-aminoethyl)amino]acetic acid
Molecular Weight (g/mol): 118.14
Molecular Formula: C4H10N2O2
InChi Key: PIINGYXNCHTJTF-UHFFFAOYSA-N
SMILES: NCCNCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)