missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Aminoethyl)glycine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 143°C |
| Color | White |
| Packaging | Glass Bottle with Plastic Insert |
| Quantity | 100 mg |
| Formula Weight | 118.14 |
| Percent Purity | ≥97.0% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(2-Aminoethyl)glycine |
Safety and Handling
| TSCA | No |