missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-valine 98.0+%, TCI America™

Catalog No. D10431G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10431G 1 g
D1043100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10431G Supplier TCI America Supplier No. D10431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1694-97-9
Molecular Formula C11H13N3O6
Molecular Weight (g/mol) 283.24
MDL Number MFCD00038096
InChI Key AYLCDVYHZOZQKM-JTQLQIEISA-N
Synonym N-Dnp-L-valine, Dnp-Val-OH
PubChem CID 96810
SMILES CC(C)C(C(=O)O)NC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 131°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 283.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-valine