accessibility menu, dialog, popup

UserName

CAS RN 1694-97-9

H3CCH3HNNOONOOOHO

CAS RN 1694-97-9

IUPAC Name: 2-[(2,4-dinitrophenyl)amino]-3-methylbutanoic acid
Synonyms: N-(2,4-dinitrophenyl)valine
Molecular Weight (g/mol): 283.24
Molecular Formula: C11H13N3O6
InChi Key: AYLCDVYHZOZQKM-UHFFFAOYNA-N
SMILES: CC(C)C(NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results