missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-proline 98.0+%, TCI America™

Catalog No. D10361G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10361G 1 g
D1036100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10361G Supplier TCI America Supplier No. D10361G

Chemical Identifiers

CAS 1655-55-6
Molecular Formula C11H11N3O6
Molecular Weight (g/mol) 281.22
MDL Number MFCD00037319,MFCD03029201,MFCD09757513
InChI Key MVZXUWLTGGBNHL-UHFFFAOYNA-N
Synonym N-Dnp-L-proline, Dnp-Pro-OH
PubChem CID 96807
ChEBI CHEBI:53086
IUPAC Name 1-(2,4-dinitrophenyl)pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 140°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 281.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-proline