missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-alanine Methyl Ester 98.0+%, TCI America™

Catalog No. D21351G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21351G Supplier TCI America Supplier No. D21351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10420-63-0
Molecular Formula C10H11N3O6
Molecular Weight (g/mol) 269.213
MDL Number MFCD00051742
InChI Key RXDDNUKGTCKLDT-LURJTMIESA-N
Synonym N-Dnp-L-alanine Methyl Ester, Dnp-Ala-OMe
PubChem CID 18642980
IUPAC Name methyl (2S)-2-(2,4-dinitroanilino)propanoate
SMILES CC(C(=O)OC)NC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 82°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 269.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-alanine Methyl Ester