missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid 95.0+%, TCI America™

Catalog No. D22541G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22541G 1 g
D2254100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22541G Supplier TCI America Supplier No. D22541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10466-72-5
Molecular Formula C12H15N3O6
Molecular Weight (g/mol) 297.27
MDL Number MFCD00038470
InChI Key ZYUWUKIAUDIXCQ-UHFFFAOYSA-N
Synonym N-Dnp-6-aminohexanoic Acid
PubChem CID 96812
ChEBI CHEBI:53698
IUPAC Name 6-[(2,4-dinitrophenyl)amino]hexanoic acid
SMILES OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 133°C
Color Yellow
Quantity 1 g
Formula Weight 297.27
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid
TSCA No
Recommended Storage Freezer