missing translation for 'onlineSavingsMsg'
Learn More

N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane 90.0+%, TCI America™

Catalog No. B406225MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B406225MG 25 mg
B4062100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B406225MG Supplier TCI America Supplier No. B406225MG
Only null left

Chemical Identifiers

CAS 1263166-93-3
Molecular Formula C17H28N2O4
Molecular Weight (g/mol) 324.421
MDL Number MFCD19705418
InChI Key YZGOWXGENSKDSE-XYPWUTKMSA-N
Synonym BCN-POE3-NH2, BCN-amine
PubChem CID 75412389
IUPAC Name [(1S,8R)-9-bicyclo[6.1.0]non-4-ynyl]methyl N-[2-[2-(2-aminoethoxy)ethoxy]ethyl]carbamate
SMILES C1CC2C(C2COC(=O)NCCOCCOCCN)CCC#C1

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 324.42
Percent Purity ≥90.0% (HPLC)
Physical Form Liquid
Chemical Name or Material N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane
TSCA No
Recommended Storage Freezer (-20 °C)