accessibility menu, dialog, popup

UserName

OOHNOONH2

CAS RN 1263166-93-3

IUPAC Name: {bicyclo[6.1.0]non-4-yn-9-yl}methyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Synonyms: bicyclo[6.1.0]non-4-yn-9-ylmethyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Molecular Weight (g/mol): 324.42
Molecular Formula: C17H28N2O4
InChi Key: YZGOWXGENSKDSE-UHFFFAOYNA-N
SMILES: NCCOCCOCCNC(=O)OCC1C2CCC#CCCC12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results