missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, Reagents

Catalog No. p-200021047
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337161 10 g
01337160 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337161 Supplier Labchem Inc Supplier No. C225330010C1
Only null left

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym N1-(Naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

Boiling Range 371°C
Color White to Pink
Boiling Point 371°C
Packaging Glass Bottle
Quantity 10 g
Specific Gravity 1.1
Grade Reagent
Physical Form Solid
Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine <br />Dihydrochloride