Learn More
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS
Used for colorimetric determination of sulfonamide in blood | CAS: 1465-25-4 | C12H16Cl2N2 | 259.174 g/mol
Supplier: Thermo Scientific Chemicals J6321414
Description
Used for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
Solubility
Soluble in water or 95% ethanol.Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.
Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
Specifications
| N-(1-Naphthyl)ethylenediamine dihydrochloride | |
| 191°C to 199°C | |
| C12H16Cl2N2 | |
| 25 g | |
| Hygroscopic; Light sensitive | |
| n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
| MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
| N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
| 15106 | |
| 259.18 | |
| Powder |
| 1465-25-4 | |
| Cream to White or Gray | |
| MFCD00012556 | |
| 3707471 | |
| 14,6409 | |
| Soluble in water or 95% ethanol;Soluble in hot water,alcohol,dilute hydrochloric acid and acetone. | |
| C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
| 259.174 | |
| CHEBI:53452 | |
| ACS |
Chemical Identifiers
| 1465-25-4 | |
| 259.174 | |
| MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
| 15106 | |
| N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
| C12H16Cl2N2 | |
| MFCD00012556 | |
| n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
| CHEBI:53452 | |
| C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-981-2
RTECSNumber : KV5330000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only