missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1-Naphthyl)ethylenediamine Dihydrochloride, ACS, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Grade | ACS |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | N-(1-Naphthyl)ethylenediamine Dihydrochloride |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.