missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent

Catalog No. AC423995000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423995000 500 g Glass bottle
AC423990050 5 g Glass bottle
AC423990250 25 g Glass bottle
AC423991000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423995000 Supplier Thermo Scientific Chemicals Supplier No. 423995000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, C12H14N2.2ClH, CAS Number-1465-25-4 | CAS: 1465-25-4 | C12H16Cl2N2 | 259.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1465-25-4
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452

Specifications

CAS Min % 98.0
CAS Max % 100.0
Decomposition Information 200°C
Melting Point 200.0°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 98% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Molecular Formula C12H14N2·2ClH
Linear Formula C10H7NHCH2CH2NH2·2HCl
Quantity 500 g
Beilstein 12, II, 699
Sensitivity (to sulfanilamide) Passes
Merck Index 15, 6491
Solubility Information Passes test
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.17
Formula Weight 259.17
Percent Purity 98+%
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material N-(1-Naphthyl)ethylenediamine, dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 215-981-2

RUO – Research Use Only