missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%

CAS: 1465-25-4 | C12H16Cl2N2 | 259.174 g/mol

Supplier:  Thermo Scientific Chemicals A1716418

 View more versions of this product

Catalog No. AAA1716418

Only null left
Explore available promotions

Description

Description

N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

Solubility
Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.

Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
Specifications

Specifications

N-(1-Naphthyl)ethylenediamine dihydrochloride
191°C to 199°C
MFCD00012556
3707471
14,6409
Soluble in hot water,alcohol,dilute hydrochloric acid and acetone.
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
259.174
CHEBI:53452
96%
1465-25-4
C12H16Cl2N2
50 g
Hygroscopic; Light sensitive
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
MZNYWPRCVDMOJG-UHFFFAOYSA-N
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
15106
259.18

Chemical Identifiers

1465-25-4
259.174
MZNYWPRCVDMOJG-UHFFFAOYSA-N
15106
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
C12H16Cl2N2
MFCD00012556
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
CHEBI:53452
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-981-2

RTECSNumber : KV5330000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only