missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine Dihydrochloride, 95%, Spectrum™ Chemical

Catalog No. p-7157882
Change view
Click to view available options
Quantity:
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604707 100 g Amber Glass Bottle
18604708 1 kg Poly Pail
18604709 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604707 Supplier Spectrum Chemical Mfg Cor Supplier No. N1031100GM
Only null left

N1031, 1465-25-4, C12H14N2·2HCl

Spectrum™ Chemical N-(1-Naphthyl)ethylenediamine Dihydrochloride

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 95%
Grade Reagent