missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Isopropoxyethyl)acetamide 93.0+%, TCI America™

Catalog No. I044125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I044125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I044125G Supplier TCI America Supplier No. I044125G

Chemical Identifiers

CAS 115910-75-3
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00191526
InChI Key PUMLTXLDRDSMGQ-UHFFFAOYSA-N
PubChem CID 14204909
IUPAC Name N-(1-propan-2-yloxyethyl)acetamide
SMILES CC(C)OC(C)NC(=O)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 145.20
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(1-Isopropoxyethyl)acetamide