missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Adamantyl)ethylenediamine 97.0+%, TCI America™

Catalog No. A14921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14921G Supplier TCI America Supplier No. A14921G
Only null left

Chemical Identifiers

CAS 37818-93-2
Molecular Formula C12H22N2
Molecular Weight (g/mol) 194.32
MDL Number MFCD02093421
InChI Key IQMUFNISQFPZJC-UHFFFAOYSA-N
Synonym 1-[N-(2-Aminoethyl)amino]adamantane
PubChem CID 303798
IUPAC Name N1-(adamantan-1-yl)ethane-1,2-diamine
SMILES NCCNC12CC3CC(CC(C3)C1)C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 194.32
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(1-Adamantyl)ethylenediamine