accessibility menu, dialog, popup

UserName

H2NNH

CAS RN 37818-93-2

IUPAC Name: N1-(adamantan-1-yl)ethane-1,2-diamine
Synonyms: N1-(adamantan-1-yl)ethane-1,2-diamine
Molecular Weight (g/mol): 194.32
Molecular Formula: C12H22N2
InChi Key: IQMUFNISQFPZJC-UHFFFAOYSA-N
SMILES: NCCNC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results