missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel 96.0+%, TCI America™
$59.65 - $59.65
Chemical Identifiers
| CAS | 264921-97-3 |
|---|---|
| Molecular Formula | C16H13N3NiO3 |
| Molecular Weight (g/mol) | 353.991 |
| InChI Key | SNANRKUUXQKAKV-UHFFFAOYSA-L |
| PubChem CID | 53384302 |
| IUPAC Name | nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate |
| SMILES | CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1738100MG
|
TCI America
P1738100MG |
100 mg |
Each for $59.65
|
|
|||||
Chemical Identifiers
| 264921-97-3 | |
| 353.991 | |
| 53384302 | |
| CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2] |
| C16H13N3NiO3 | |
| SNANRKUUXQKAKV-UHFFFAOYSA-L | |
| nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate |
Specifications
| 264921-97-3 | |
| 100 mg | |
| CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2] | |
| 353.991 | |
| 353.99 | |
| Crystalline Powder |
| C16H13N3NiO3 | |
| SNANRKUUXQKAKV-UHFFFAOYSA-L | |
| nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate | |
| 53384302 | |
| ≥96.0% (T) | |
| [N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel |
Safety and Handling
TSCA : No