accessibility menu, dialog, popup

UserName

CAS RN 264921-97-3

H3CNOONNONi(E/Z)(E/Z)

CAS RN 264921-97-3

IUPAC Name: nickel(2+) 2-{[1-(2-{[oxido(pyridin-2-yl)methylidene]amino}phenyl)ethylidene]amino}acetate
Synonyms: nickel(2+) 2-{[1-(2-{[oxido(pyridin-2-yl)methylidene]amino}phenyl)ethylidene]amino}acetate
Molecular Weight (g/mol): 353.99
Molecular Formula: C16H13N3NiO3
InChi Key: SNANRKUUXQKAKV-UHFFFAOYSA-L
SMILES: [Ni++].CC(=NCC([O-])=O)C1=CC=CC=C1N=C([O-])C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results