missing translation for 'onlineSavingsMsg'
Learn More

[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel 96.0+%, TCI America™

Catalog No. P1738100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1738100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1738100MG Supplier TCI America Supplier No. P1738100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 264921-97-3
Molecular Formula C16H13N3NiO3
Molecular Weight (g/mol) 353.991
InChI Key SNANRKUUXQKAKV-UHFFFAOYSA-L
PubChem CID 53384302
IUPAC Name nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate
SMILES CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2]

Specifications

Quantity 100 mg
Formula Weight 353.99
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel