missing translation for 'onlineSavingsMsg'
Learn More

N-(1,1,3,3-Tetramethylbutyl)formamide 98.0+%, TCI America™

Catalog No. T17585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17585G Supplier TCI America Supplier No. T17585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10151-02-7
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00039569
InChI Key NBXAPOVFSGOATB-UHFFFAOYSA-N
Synonym N-Formyl-tert-octylamine, N-Formyl-1,1,3,3-tetramethylbutylamine, tert-Octylformamide
PubChem CID 82410
IUPAC Name N-(2,4,4-trimethylpentan-2-yl)formamide
SMILES CC(C)(C)CC(C)(C)NC=O

Specifications

Color Colorless
Boiling Point 110°C
Quantity 5 g
Formula Weight 157.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(1,1,3,3-Tetramethylbutyl)formamide