accessibility menu, dialog, popup

UserName

CAS RN 10151-02-7

IUPAC Name: N-(2,4,4-trimethylpentan-2-yl)formamide
Synonyms: N-(2,4,4-trimethylpentan-2-yl)formamide
Molecular Weight (g/mol): 157.26
Molecular Formula: C9H19NO
InChi Key: NBXAPOVFSGOATB-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)NC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results