missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MZT2B Polyclonal Antibody

Description
Immunogen sequence: MAAQGVGPGP GSAAPPGLEA ARQKLALRRK KVLSTEEMEL YELAQAAGGA IDPDVFKILV DLLKLNVAPL AVFQMLKSMC AGQRLASEPQ DPAAVSLPTS SVPETRGRNK GSAALGGALA LAERSSREGS SQRMPRQPSA TRLPKGGGPG KSPTRGST; Positive Samples: U-87MG, Jurkat, HeLa, MCF-7, 293T; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle.
MZT2B (Mitotic Spindle Organizing Protein 2B) is a Protein Coding gene. An important paralog of this gene is MZT2A.
Specifications
Specifications
| Antigen | MZT2B |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.91 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MZT2B |
| Gene Accession No. | Q6NZ67 |
| Gene Alias | FAM128B; family with sequence similarity 128, member B; mitotic spindle organizing protein 2B; mitotic-spindle organizing protein 2B; Mitotic-spindle organizing protein associated with a ring of gamma-tubulin 2B; MOZART2B; MZT2B |
| Gene Symbols | MZT2B |
| Show More |