missing translation for 'onlineSavingsMsg'
Learn More

Myristoyl Chloride 98.0+%, TCI America™

Catalog No. T008625G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T008625G 25 g
T0086400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T008625G Supplier TCI America Supplier No. T008625G

Chemical Identifiers

CAS 112-64-1
Molecular Formula C14H27ClO
Molecular Weight (g/mol) 246.819
MDL Number MFCD00000741
InChI Key LPWCRLGKYWVLHQ-UHFFFAOYSA-N
Synonym myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r
PubChem CID 66986
IUPAC Name tetradecanoyl chloride
SMILES CCCCCCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
UN Number 3265
Formula Weight 246.82
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Myristoyl Chloride
EINECSNumber (2)-0632
TSCA Yes