accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 112-64-1

IUPAC Name: tetradecanoyl chloride
Synonyms: tetradecanoyl chloride
Molecular Weight (g/mol): 246.82
Molecular Formula: C14H27ClO
InChi Key: LPWCRLGKYWVLHQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(Cl)=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)