missing translation for 'onlineSavingsMsg'
Learn More

Myristoyl chloride, 97%

CAS: 112-64-1 | C14H27ClO | 246.82 g/mol

$160.42 - $160.42

Chemical Identifiers

CAS 112-64-1
Molecular Formula C14H27ClO
Molecular Weight (g/mol) 246.82
MDL Number MFCD00000741
InChI Key LPWCRLGKYWVLHQ-UHFFFAOYSA-N
Synonym myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r
PubChem CID 66986
IUPAC Name tetradecanoyl chloride
SMILES CCCCCCCCCCCCCC(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC180631000
SDS View Documents
Thermo Scientific Chemicals
180631000
100 g Glass bottle
Each for $160.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-64-1
246.82
LPWCRLGKYWVLHQ-UHFFFAOYSA-N
66986
CCCCCCCCCCCCCC(=O)Cl
C14H27ClO
MFCD00000741
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r
tetradecanoyl chloride

Specifications

112-64-1
100.0
172°C to 176°C (5.0 mbar)
Authentic
Glass bottle
1.4480 to 1.45
MFCD00000741
02, 368
Solubility in water: may decompose
CCCCCCCCCCCCCC(=O)Cl
246.82
246.82
Liquid
96.0
-1°C
>100°C
96% min. (GC)
C14H27ClO
CH3(CH2)12COCl
100 g
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r
LPWCRLGKYWVLHQ-UHFFFAOYSA-N
tetradecanoyl chloride
66986
97%
Myristoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May form explosive peroxides.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 203-994-6

SDS
Documents

Documents

RUO – Research Use Only