missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myristicin, ≥97% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 607-91-0 |
| Linear Formula | C11H12O3 |
| MDL Number | MFCD00133549 |
| Synonym | 5-Allyl-2,3-(methylendioxy)anisole; 6-Allyl-4-methoxy-1,3-benzodioxole |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 192.21 |
| Percent Purity | ≥97% (GC) |
| Grade | Analytical Standard |