missing translation for 'onlineSavingsMsg'
Learn More

Myristanilide 99.0+%, TCI America™

Catalog No. p-7132317
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M071825G 25 g
M0718500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M071825G Supplier TCI America Supplier No. M071825G
Only null left

Chemical Identifiers

CAS 622-56-0
Molecular Formula C20H33NO
Molecular Weight (g/mol) 303.49
MDL Number MFCD00059285
InChI Key LYSJUAWWUBVYCB-UHFFFAOYSA-N
Synonym myristanilide, tetradecanamide, n-phenyl, n-phenyltetradecanamid, n-myristoylaniline, n-phenylmyristamide, myristic acid anilide, tetradecanoic acid phenylamide
PubChem CID 69324
IUPAC Name N-phenyltetradecanamide
SMILES CCCCCCCCCCCCCC(=O)NC1=CC=CC=C1

Specifications

Melting Point 84°C
Color White
Boiling Point 113°C
Quantity 25 g
Formula Weight 303.49
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Myristanilide
EINECSNumber (3)-2698
TSCA Yes