accessibility menu, dialog, popup

UserName

H3COHN

CAS RN 622-56-0

IUPAC Name: N-phenyltetradecanamide
Synonyms: N-phenyltetradecanamide
Molecular Weight (g/mol): 303.49
Molecular Formula: C20H33NO
InChi Key: LYSJUAWWUBVYCB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results