missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myrcene, Analytical Standard, ≥90.0% (GC), MilliporeSigma™ Supelco™
Myrcene is an acyclic monoterpene having antimutagenic properties, which can be found in the essential oils of many useful plants like lemongrass, hop, bay, verbena and also others.
$113.22 - $453.91
Specifications
| Packaging | Bottomless Glass Bottle with Fused Cone |
|---|---|
| CAS | 123-35-3 |
| Vapor Pressure | ˜7 mmHg (20°C) |
| Vapor Density | 4.7 (vs. air) |
| Density | 0.791 g/mL (at 25°C) |
Specifications
| Bottomless Glass Bottle with Fused Cone | |
| ˜7 mmHg (20°C) | |
| 0.791 g/mL (at 25°C) | |
| H2C=CHC(=CH2)CH2CH2CH=C(CH3)2 | |
| 167°C | |
| 136.23 | |
| Analytical Standard |
| 123-35-3 | |
| 4.7 (vs. air) | |
| Neat | |
| MFCD00008908 | |
| beta-Myrcene; 7-Methyl-3-methylene-1,6-octadiene | |
| ≥90.0% (GC) |