missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYPN Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5149848

Catalog No. PA5149848

Only null left
Explore available promotions

Description

Description

MYPN Polyclonal Antibody for Western Blot, ICC/IF, IHC

Striated muscle in vertebrates comprises large proteins which must be organized properly to contract efficiently. Z-lines in striated muscle are a sign of this organization, representing the ends of actin thin filaments, titin, nebulin or nebulette and accessory proteins required for structure and function. This gene encodes a protein which interacts with nebulin in skeletal muscle or nebulette in cardiac muscle and alpha-actinin. In addition, this gene product can interact with a protein with the I-band indicating it has a regulatory as well as structural function. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

MYPN
Polyclonal
Unconjugated
MYPN
1110056A04Rik; 145 kDa sarcomeric protein; 9830112J03; AI853556; CMD1DD; CMH22; Kiaa4170; mKIAA4170; MYOP; Myopalladin; MYPN; RCM4; sarcomeric protein myopalladin, 145 kDa (MYOP); wu:fc17a02
Rabbit
Protein A
RUO
100624901, 309760, 68802, 84665
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q5DTJ9, Q86TC9
MYPN
Recombinant Myopalladin (MYPN) (Pro945-Leu1320).
200 μL
Primary
Human, Mouse, Rat, Pig
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions