missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MyPIPETMANTM Select Single Channel, Metal Ejector
GREENER_CHOICE

Catalog No. FP10004STG
Change view
Click to view available options
Model:
P100
P1000
P200
Volume (Metric):
10 to 100 μL
100 to 1000 μL
20 to 200 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Volume (Metric)
FP10004STG P100 10 to 100 μL
FP10005STG P200 20 to 200 μL
FP10006STG P1000 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FP10004STG Supplier Gilson™ Supplier No. FP10004ST

Ergonomic, safe, and reliable, MyPIPETMANTM Select offers volume setting choice and optional handle personalization for optimal use in various protocols.

MyPIPETMANTM Select fully-autoclavable, air-displacement pipettes cover a full volume range, including:

  • Variable volume single channel models covering a volume range from 0.2 μL to 10 mL
  • Variable volume multichannel models covering a volume range from 0.5 μL to 300 μL (8 and 12-channel models)

Set and Secure the Volume your Preferred Way

  • Simply adjust the volume by turning the push button of the pipette with the unique, patented TrilockTM volume-locking system.
    • You can set the volume in your preferred way:
      • Free volume setting position for quick and effortless volume adjustment
      • Light ratchet volume setting for precise volume adjustment
      • Volume locked position to secure pipetting

Ergonomic Design

  • Lightweight and well-balanced, MyPIPETMANTM Select has an elegant design with a textured effect for a better grip.
  • With no compromise on quality and performance, the built-in ejection system with an integrated lever provides reduced tip ejection forces compared to direct ejection systems.
  • This patented ejection system reinforces the pipette's ergonomics for convenient use throughout the day.

Personalize your Pipette for an Optimal Use

  • MyPIPETMANTM Select has optional handle printing on demand. Identify your pipette to avoid any risk of contamination by dedicating it to a specific liquid or workflow step and improve your lab's Lean Management process.

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Tan
Disposable Reusable
For Use With D-200, DF100 Tip
Grip Type Ergonomic, Textured
Material Polypropylene, PVDF, PBT
Model P100
Number of Channels 1
Product Line MyPipetman
Programmability No
Sterility Non-sterile
Type Manual Air Displacement Pipette
Volume (Metric) 10 to 100 μL
Quantity 1
Width (English) 4.25 in
Fit for Purpose Certified Quality
Show More Show Less