missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MyPIPETMAN™ Select 4 Pipette Kit PROMO

Catalog No. 01257530W25
Change view
Click to view available options
Gilson™ Select 4 Pipette Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01257530W25 Gilson™ Select 4 Pipette Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257530W25 Supplier Gilson™ Supplier No. F167390PM

Kit includes pipettes with reinforced ergonomics for optimal use in many protocols and most liquid handling tasks in molecular biology, immunology and cell culture. Included are the most commonly used pipettes: P2, P20, P200, and P1000; plus tips and accessories.

MyPIPETMAN™ Select 4-Pipette Kit includes extra customization accessories (colored windows) are also provided to identify the use of the pipettes for a specific liquid, room, or technique.

  • Simply adjust the volume by turning the push button of the pipette with the unique, patented Trilock™ volume-locking system
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
  • Lightweight and well-balanced, MyPIPETMAN™ Select has an elegant design and an optimized grip
  • Built-in ejection system provides reduced tip ejection forces for convenient use throughout the day
  • Fully autoclavable without disassembly for maximum convenience and protection from cross-contamination
  • Autoclavable plastic tip ejectors, as extra accessories, allow using the pipettes when handling corrosive liquids
  • Pipettes have a longer life span and can be used with different kinds of liquids without any risk of damaging the metal tip ejectors
  • SINGLE™ pipette holders are also provided in each kit to store each pipette vertically on a wall, protecting the pipette from contamination without taking up any precious bench space
  • Includes: 4 MyPIPETMAN™ Select pipettes: P2, P20, P200, and P1000 equipped with metal ejectors, 3 PIPETMAN™ DIAMOND Tips TIPACKS (models: DL10, D200 and D1000), 4 SINGLE pipette holders, 4 white plastic tip ejectors, 6 colored windows (red, yellow, blue, green, white, and dark blue)

Specifications

Compatible Tips Yes
Autoclavable Autoclavable
Color Blue/White
Disposable Reusable
For Use With Universal Pipette Tip
Grip Type Ergonomic
Material Polypropylene, Stainless Steel
Model P2, P20, P200, P1000
Number of Channels 1
Product Line Pipetman
Sterility Non-sterile
Type Pipette Kit
Volume (Metric) 1000 μL
Weight (Metric) 1 kg
Fit for Purpose PCR Ready
Show More Show Less