Learn More
Gilson™ MyPIPETMANTM Enterprise Single Channel Pipettes
GREENER_CHOICE
The Enterprise pipettes combine reliability, ergonomics, and personalization options to ensure efficiency and answer your ranging pipetting needs.
Supplier:  Gilson™ FP10002E
 View more versions of this product
Catalog No. FP10002EG
Only null left
Description
Description
MyPIPETMANTM Enterprise pipettes are produced in a series of at least 150 units (in a mix of different volumes). All pipettes inside a series have the same technical features, design, and information printed on the handle.
  • Select your preferred technical features to optimize the use of your pipette and overall productivity in the lab
  • Color choices and printing options on your pipette allow for better organization and identification within your workflow
Specifications
Chemical Identifiers
Diamond Tips, Amplipur Tips
Autoclavable
Reusable
Polypropylene, PVDF, PBT
MyPipetman
Non-sterile
1
No
Assorted
Textured
1
No
Mechanical Pipette
4.25 in
Specifications
Diamond Tips, Amplipur Tips
Autoclavable
Reusable
Textured
P10
MyPipetman
Non-sterile
10 μL
4.25 in
No
Assorted
Pipette Tip 0.5 to 20 μL
Polypropylene, PVDF, PBT
1
No
Mechanical Pipette
1
Product Suggestions
Product Suggestions