missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myotilin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529059 Supplier Invitrogen™ Supplier No. PA529059
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (93%), Rat (94%), Pig (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.

Specifications

Antigen Myotilin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MYOT
Gene Accession No. Q9UBF9
Gene Alias 5530402I04Rik; 57 kDa cytoskeletal protein; LGMD1; LGMD1A; MFM3; Myo; Myofibrillar titin-like Ig domains protein; MYOT; Myotilin; titan-like protein; titin immunoglobulin domain protein; titin immunoglobulin domain protein (myotilin); Ttid; TTOD
Gene Symbols MYOT
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Myotilin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9499
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less