missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myosin heavy chain 1 Recombinant Rabbit Monoclonal Antibody (PSH05-12)

Catalog No. MA555602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA555602 Supplier Invitrogen™ Supplier No. MA555602
Only null left

Rabbit Recombinant Monoclonal Antibody

Positive Control: Human skeletal muscle tissue, mouse skeletal muscle tissue, rat skeletal muscle tissue. Tissue Specificity: Tissue enriched (skeletal). Subcellular Location: Cytoplasm, myofibril. Sequence Similarities: 100% Mouse/Rat. Predicted band size: 223 kDa.

Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).

Specifications

Antigen Myosin heavy chain 1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PSH05-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Myh1
Gene Accession No. P12882, Q5SX40
Gene Alias A530084A17Rik; epididymis luminal protein 71; HEL71; IId; IId/x; MdMs; MHC2X/D; MHC-2X/D; MYH1; MYHa; MYHC; myHC-2x; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; myosin heavy chain 1; Myosin heavy chain 2x; myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin-1; type 2X myosin heavy chain
Gene Symbols Myh1
Host Species Rabbit
Immunogen Synthetic peptide within human MYH1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17879, 287408, 4619
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less