missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myogenin Recombinant Rabbit Monoclonal Antibody (JE60-39)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA555604

Catalog No. MA555604


Only null left

Description

Description

Positive Control: Differentiated C2C12 cell lysate. Tissue Specificity: Group enriched (skeletal muscle, tongue). Subcellular Location: Nucleus. Sequence Similarities: 100% Mouse/Rat. Predicted band size: 25 kDa.

Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
Specifications

Chemical Identifiers

Myogenin
1 mg/mL
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P12979, P15173, P20428
17928, 29148, 4656
Synthetic peptide within human myogenin aa 1-50/224.
Antibody
JE60-39
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
MYOG
bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4)
MYOG
Primary
Protein A

Specifications

Myogenin
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P12979, P15173, P20428
MYOG
Synthetic peptide within human myogenin aa 1-50/224.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry
JE60-39
Unconjugated
MYOG
bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4)
Rabbit
Protein A
RUO
17928, 29148, 4656
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions