missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150253
Description
MYO1D Polyclonal Antibody for Western Blot, ICC/IF, IHC
Unconventional myosin that functions as actin-based motor protein with ATPase activity. Plays a role in endosomal protein trafficking, and especially in the transfer of cargo proteins from early to recycling endosomes. Required for normal planar cell polarity in ciliated tracheal cells, for normal rotational polarity of cilia, and for coordinated, unidirectional ciliary movement in the trachea. Required for normal, polarized cilia organization in brain ependymal epithelial cells.Chemical Identifiers
| MYO1D | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O94832, Q5SYD0, Q63357 | |
| 25485, 338367, 4642 | |
| Recombinant Myosin ID (MYO1D) (Ser581-Glu847). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MYO1D | |
| 9930104H07Rik; AW544947; D11Ertd9e; KIAA0727; Myo1c; MYO1D; myosin 1C; myosin heavy chain myr 4; myosin IC; myosin ID; myosin1C; myosin-1d; myosin-I gamma; myosin-Id; Myr2; Myr4; PPP1R108; protein phosphatase 1, regulatory subunit 108; Unconventional myosin from rat 4 for myosin I heavy chain; unconventional myosin-Id | |
| MYO1D | |
| Primary | |
| Protein A |
Specifications
| MYO1D | |
| Polyclonal | |
| Unconjugated | |
| MYO1D | |
| 9930104H07Rik; AW544947; D11Ertd9e; KIAA0727; Myo1c; MYO1D; myosin 1C; myosin heavy chain myr 4; myosin IC; myosin ID; myosin1C; myosin-1d; myosin-I gamma; myosin-Id; Myr2; Myr4; PPP1R108; protein phosphatase 1, regulatory subunit 108; Unconventional myosin from rat 4 for myosin I heavy chain; unconventional myosin-Id | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25485, 338367, 4642 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O94832, Q5SYD0, Q63357 | |
| MYO1D | |
| Recombinant Myosin ID (MYO1D) (Ser581-Glu847). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |