missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol 99.0+%, TCI America™

Catalog No. p-7131174
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
I004025G 25 g
I0040500G 500 g
2 options

Catalog No. I004025G

Supplier: TCI America I004025G

Only null left

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 226°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material myo-Inositol
EINECSNumber (3)-3265&(8)-0520
RTECSNumber NM7520800
TSCA Yes