missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol, 99%

Catalog No. p-7041954
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6082822 100 g
AAJ6082836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6082822 Supplier Thermo Scientific Chemicals Supplier No. J6082822
Only null left

A lipotropic agent | CAS: 87-89-8 | C6H12O6 | 180.156 g/mol

A lipotropic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 222°C to 227°C
Color White
Odor Odorless
Quantity 100 g
Beilstein 1907329
Merck Index 14,4978
Formula Weight 180.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material myo-Inositol
EINECSNumber 201-781-2
RTECSNumber NM7520800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only