missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYLK (MLCK), GST Tag Recombinant Protein

Catalog No. PV4339
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4339 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4339 Supplier Invitrogen™ Supplier No. PV4339
Only null left

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic

MYLK (MLCK) is a serine/threonine kinase that phosphorylates myosin regulatory light chains to assist in the production of contractile activity (myosin-actin interaction). MLCK is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.

Specifications

Accession Number Q15746
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 4638
Molecular Weight (g/mol) 67.3 kDa
Name Human MYLK (MLCK), GST Tag
Quantity 10 μg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias 155 kda myosin light chain kinase homolog; 9530072E15Rik; A clone isolated from a chicken gizzard cDNA library which has high homology with the 3'-end sequence of the chicken gizzard myosin light chain kinase (MLCK) cDNA; The 5'-leader seqeuence derives from the intronic sequence of the MLCK gene.; A930019C19Rik; AAT7; AW489456; kinase related protein; kinase-related protein; KRP; KRP/Telokin; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; mylk; MYLK1; myosin light chain kinase; myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; nmMlck; smMLCK; smooth muscle myosin light chain kinase; smooth muscle myosin light chain kinase (61-kDa active fragment); smooth muscle myosin light chain kinase precursor (EC 2.7.2.37); telokin; unnamed protein product
Common Name MYLK
Kinase Family MLCK
Protein Length 1428-1771
Product Type MYLK (MLCK) Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinase and inhibitors
Form Liquid
Protein Subtype Serine/threonine kinase
Show More Show Less