missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121254
Description
Positive test controls include: SKOV3, 22Rv1, Mouse brain, Mouse small intestine, Rat brain, Rat testis. Immunogen sequence: MPPKKDVPVK KPAGPSISKP AAKPAAAGAP PAKTKAEPAV PQAPQKTQEP PVDLSKVVIE FNKDQLEEFK EAFELFDRVG DGKILYSQCG DVMRALGQNP TNAEVLKVLG NPKSDELKSR RVDFETFLPM LQAVAKNRGQ GTYEDYLEGF RVFDKEGNGK VMGAELRHVL TTLGEKMTEE EVETVLAGHE DSNGCINYEA FLKHILSV.
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue.Specifications
| MYL6B | |
| Polyclonal | |
| Unconjugated | |
| Myl6b | |
| 5730437E04Rik; BC037527; MLC1SA; MYL6B; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; myosin light chain 6B; myosin, light chain 6B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6B; myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6B; smooth muscle and nonmuscle myosin light chain alkali 6B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140465, 216459, 685883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.68 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14649, Q8CI43 | |
| Myl6b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MYL6B (NP_002466.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |