missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119955
Description
Positive Control: Mouse lung tissue lysate, human heart tissue lysate, rat lung tissue, rat heart tissue. Subcellular Location: Myosin complex, cytosol, A band.
Regulatory light chain of myosin. Does not bind calcium.Specifications
| MYL4 | |
| Polyclonal | |
| Unconjugated | |
| MYL4 | |
| ALC1; AMLC; ELC; ELC1a; GT1; MLC1; MLC1a; MLC1EMB; MYL4; Myla; myosin light chain 1, atrial isoform; myosin light chain 1, atrial/fetal isoform; myosin light chain 1, embryonic muscle/atrial isoform; Myosin light chain 4; myosin light chain alkali GT-1 isoform; myosin light chain, alkali, cardiac atria; myosin, atrial/fetal muscle, light chain; myosin, light chain 4; myosin, light chain 4, alkali; atrial, embryonic; myosin, light polypeptide 4; myosin, light polypeptide 4, alkali; atrial, embryonic; PRO1957 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17896, 4635, 688228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P09541, P12829, P17209 | |
| MYL4 | |
| Recombinant protein within human MYL4 aa 10-197/197. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |