missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYF6 Monoclonal Antibody (5D7)
GREENER_CHOICE

Catalog No. PIMA517131
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517131 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517131 Supplier Invitrogen™ Supplier No. MA517131
Only null left

Mouse Monoclonal Antibody

MA5-17131 targets MYF6 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17131 immunogen is purified recombinant fragment of human MYF6 (amino acids: 85-150) expressed in E. Coli. MA5-17131 detects MYF6 which has a predicted molecular weight of approximately 27kDa.

Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.

Specifications

Antigen MYF6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Myf6
Gene Accession No. P23409
Gene Alias bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6
Gene Symbols Myf6
Host Species Mouse
Immunogen Purified recombinant fragment of human MYF6 (amino acids: 85-150) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less